Learn More
Medchemexpress LLC Carnosic acid | 3650-09-7 | MFCD02259459 | 332.43 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0644R
Encompass_Preferred
Carnosic acid is an orally active lipid absorption inhibitor. It has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity. It also has therapeutic potential in different types of cancer and neuroprotective effects in neurodegenerative diseases through activation of the antioxidant NRF2/ARE pathway. It is a phenolic diterpene found in the leaves of Lamiaceae plants, particularly Rosmarinus officinalis L. and Salvia officinalis.
  • Orally active lipid absorption inhibitor
  • Demonstrates inhibition of oxidative stress and inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • Potential therapeutic agent in various cancers
  • Neuroprotective effects in neurodegenerative diseases
Catalog No. 50-003-89183