Learn More
Medchemexpress LLC Carnosic acid | 3650-09-7 | MFCD02259459 | 332.43 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN064450MG
Encompass_Preferred
Carnosic acid is an orally active lipid absorption inhibitor found in the leaves of Lamiaceae plants like *Rosmarinus officinalis L.* and *Salvia officinalis*. It inhibits oxidative stress and inflammation, suppresses cell proliferation, and exhibits antibacterial activity. Carnosic acid has shown therapeutic potential in various cancers and neuroprotective effects in neurodegenerative diseases by activating the NRF2/ARE pathway.
  • Inhibits lipid absorption
  • Reduces oxidative stress and inflammation
  • Suppresses cell proliferation
  • Exhibits antibacterial activity
  • Offers therapeutic potential in cancers
  • Shows neuroprotective effects in neurodegenerative diseases
Catalog No. 50-003-88395