Learn More
Cayman Chemical Carotene 10g
SDP
Supplier:  Cayman Chemical 16837 10G
Encompass_Preferred
An internal standard for the quantification of RCS-4 N-(5-hydroxypentyl) metabolite by GC- or LC-MS intended for research and forensic applications.
Catalog No. 50-296-5936