Learn More
YMC America, Inc Carotenoid col 3um 250x2.1mm
SDP
Supplier:  YMC America, Inc CT99S0325Q1WT
Encompass_Preferred
YMC Carotenoid is a C30 bonded silica-based reversed phase column that is markedly more hydrophobic than classical C18 phases Even when pure organic eluents are applied many sample solutes are retained The YMC Carotenoid C30 stationary phase provides sufficient phase thickness to enhance interaction with long chain molecules therefore geometric and positional isomers of conjugated double-bonding systems are recognized and resolved It can be used to separate carotenoids in blood samples food products natural product extracts and commercial preparations In addition YMC Carotenoid operates with low aqueous or non-aqueous mobile phases that are desirable in LC/MS and prep fraction recovery
Catalog No. 50-008-77072