Learn More
Medchemexpress LLC Caroverine hydrochlo 100mg | 55750-05-5 | 100MG
SDP
Supplier:  Medchemexpress LLC HY106467B100MG
Encompass_Preferred
Caroverine hydrochloride is a potent competitive and reversible antagonist of NMDA and AMPA glutamate receptor Caroverine hydrochloride is also an antioxidant and calcium-blocking agent that exhibits vasorelaxant action Caroverine hydrochloride can be used for the research of inner ear tinnitus[1 [2 [3
Catalog No. 50-002-40095