Learn More
Medchemexpress LLC Caroverine hydrochloride | 55750-05-5 | MFCD01684929 | ≥98.0% | 401.93 g·mol⁻¹ | C22H28ClN3O2 | 50 MG

Supplier: Medchemexpress LLC HY106467B50MG
Caroverine hydrochloride is the analytical standard of caroverine supplied for research and analytical applications. It functions as a competitive, reversible antagonist of NMDA and AMPA glutamate receptors and also shows antioxidant and calcium-blocking (vasorelaxant) activity, making it useful for studies of glutamatergic signaling and inner ear tinnitus.
- Analytical standard intended for research and analytical use.
- Competitive, reversible antagonist of NMDA and AMPA receptors.
- Exhibits antioxidant and calcium-blocking vasorelaxant activity.
- Molecular weight 401.93 g·mol⁻¹; formula C22H28ClN3O2.
- Soluble in DMSO and water; clear solutions reported at low mg/mL concentrations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.