Learn More
Medchemexpress LLC Caroverine hydrochloride | 55750-05-5 | MFCD01684929 | 98.9% | 401.93 g/mol | C22H28ClN3O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY106467B25MG
Encompass_Preferred
Caroverine hydrochloride is a research-grade small molecule that functions as a competitive, reversible antagonist of NMDA and AMPA glutamate receptors. It also exhibits antioxidant and calcium-blocking activity with vasorelaxant effects, and is commonly used in studies of inner ear tinnitus and glutamatergic signaling.
  • Competitive, reversible NMDA and AMPA antagonist
  • Antioxidant and calcium-blocking activity
  • Used in tinnitus and glutamatergic signaling research
  • High purity (98.9%) for reproducible results
  • White to off-white solid, easy to weigh and handle
  • Available in small pack sizes suitable for laboratory use
Catalog No. 50-002-40158