Learn More
MSC Clarke Carpet Extractor, 18 in Path, 12 gal Tank, 120 PSI
Supplier:  MSC 35632389
Encompass
This self-contained carpet extractor features an 18-inch cleaning path and a 12-gallon tank capacity. It is equipped with a 1.5 hp motor and delivers 120 PSI pump pressure for effective carpet cleaning. The unit also includes a 10-gallon recovery tank capacity and is designed to be portable.
  • 1.5 hp motor.
  • 12 gal solution tank capacity.
  • 10 gal recovery tank capacity.
  • 120 PSI pump pressure.
  • 18 in cleaning width.
  • Portable design.
  • Front-mounted drain hose.
  • Self-aligning vacuum shoe.
  • One touch control panel.
  • Removable tank.
  • Adjustable handle.
  • Floating brush head.
  • ETL certified.
Catalog No. 50-283-6384
Add to Cart
Request a Quote