Learn More
Medchemexpress LLC κ-Carrageenan | 11114-20-8 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY138962
Encompass_Preferred
κ-Carrageenan is a natural polymer predominantly found in red seaweeds. It functions as an effective agent carrier for delivering curcumin into cancer cells to induce apoptosis. Additionally, κ-carrageenan can act as a potential inflammatory agent that exacerbates existing intestinal inflammation.
  • Natural polymer predominantly available in red seaweeds.
  • Effective agent carrier to deliver curcumin in cancer cells and to induce apoptosis.
  • Serves as a potential inflammatory agent that magnifies existing intestinal inflammation.
  • Used in animal modeling to create paw edema, colitis, and thrombosis models.
  • Exhibits oral activity with rapid absorption, slow elimination, and prolonged retention in the body in mice.
  • Accumulates in the liver and kidneys.
Catalog No. 50-003-51583