Learn More
Medchemexpress LLC Carrageenan | 9000-07-1 | Not applicable (polymeric polysaccharide; no single chemical formula) | 25G
SDP
Supplier:  Medchemexpress LLC HY12547425G
Encompass_Preferred
Carrageenan is a seaweed-derived sulfated polysaccharide used as a biochemical reagent in life-science research. It is supplied as a white to off-white solid (CAS 9000-07-1) in small research pack sizes and is employed as a viscous agent, thickener, or gelling component in formulation and as a proinflammatory agent in preclinical assays.
  • Seaweed-derived sulfated polysaccharide suitable for research use.
  • White to off-white solid, supplied in small research packs.
  • Useful as a viscous agent, thickener, and gelling component.
  • Used as a proinflammatory agent in basic research assays.
  • Analytical characteristics: loss on drying ~11.5%, viscosity ~15 mPa·s.
Catalog No. 50-002-36124