Learn More
Essendant Rubbermaid Commercial Square Storage Container, 12 qt, 10.5 x 11.31 x 7.75, Clear, Plastic
Supplier:  Essendant RCPFG631200CLR
Encompass_Preferred
The Rubbermaid Commercial Space Saving Food Storage Containers keep food fresh organized and ready-to-use Featuring easy-to-read graduated measurements in blue for liters and red for quarts Constructed of break-resistant polycarbonate they have a temperature range of -40 to 212 F making them perfect for everything from freezer storage to sous vide cooking Commercial dishwasher safe they feature durable handles with drain holes for proper washing and drying Certified for food contact assist in complying with HACCP guidelines Food Container Type- Base Material(s)- Polycarbonate Material Type- Plastic Interior Volume Capacity- 12 qt
Catalog No. 50-000-26150