Learn More
TrippNT CART TRIPPNT ELEMENT 03 WHITE
SDP
Supplier:  TrippNT 51498
Encompass_Preferred
TrippNT's 51498 Element 03 Cart is a superior value healthcare cart which can be customized to use as a crash cart, phlebotomy cart, ER cart, OR cart and more. This space saving slim design maximizes your work area. Constructed of 2 skins of corrosion resistant aluminum with a high density polyethylene core, this cart is fully customizable using built in tracks and has superior maneuverability with a retractable command handle. It comes standard with 4-inch nylon rubber non-marking casters, but 5 inch or 3 inch casters are available. There is plenty of room for storage; 3 full extension drawers with a large bulk storage area. The spacious flat top is a workspace on the go. 21 x 35 x 18 inches (width height depth); 47 pounds. All TrippNT Carts have a 100% Total Replacement Lifetime Guarantee.
Catalog No. 50-105-7153