Learn More
Medchemexpress LLC Carteolol hydrochloride | 51781-21-6 | 99.9% | 328.83 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY17495A200MG
Encompass_Preferred
Carteolol hydrochloride is a non-selective beta blocker used to treat glaucoma. It also acts as a beta-adrenergic antagonist with applications as an anti-arrhythmia agent, an anti-angina agent, an antihypertensive agent, and an antiglaucoma agent. It has demonstrated significant inhibition of H2O2-induced cell damage and scavenging of O2, providing protection against UVB-induced HCEC damage.
  • Used to treat glaucoma
  • Acts as a beta-adrenergic antagonist
  • Exhibits anti-arrhythmia properties
  • Functions as an anti-angina agent
  • Serves as an antihypertensive agent
  • Provides protection against UVB-induced HCEC damage
Catalog No. 50-003-86851