Learn More
Essendant CARTRIDGEMLTGAS/VPR2/PK
Supplier:  Essendant MMM6006
Encompass_Preferred
Low-profile design provides excellent field of vision Two cartridges per pack Effective against certain organic vapors and other gases Filter Type(s)- Gas Organic Vapors Filtration For- Certain Organic Vapors Other Gases For Use With- 3M Full Facepiece Series 6000 7800 FF-400 Half Facepiece Reusable Respirators Series 6000 6500 7500 Scott AV-3000 HT Facepieces AV-3000 SureSeal Facepieces Quantity- 2/Pack
Catalog No. 50-000-31768