Learn More
Teledyne ISCO, Inc REDISEP EMPTY DISPOSABLE SAMPLE LOAD CARTRIDGES, HOLDS UP TO 25 GRAM (PACKAGE OF 30)
Supplier:  Teledyne ISCO, Inc 693873240
Encompass
Polypropylene cartridges available either prepacked with normal-phase or reversed-phase sorbent, or empty, to be filled with user-supplied sorbent.
Catalog No. 50-901-8245