Learn More
Teledyne ISCO, Inc REDISEP PREPACKED SILICA GEL DISPOSABLE SAMPLE LOAD CARTRIDGES, 12 GRAM (PACKAGE OF 15)
Supplier:  Teledyne ISCO, Inc 693873243
Encompass
Sample load prepacked silica gel Universal RediSep cartridges, 12 gram, pkg 15. For use on Companion, Companion XL, Sg100c, Sq16x, Retrieve and the Torrent/Rf SLCC, 60-5237-048.
Catalog No. 50-901-8248