Learn More
Selleck Chemical LLC Carvedilol EP IMpurity E
SDP
Supplier:  Selleck Chemical LLC S572325MG
Encompass_Preferred
Carvedilol is a nonselective beta blocker/alpha-1 blocker and used in management of congestive heart failure (CHF)
Catalog No. 50-000-03145