Learn More
Medchemexpress LLC Carvedilol phosphate hemihydrate | 610309-89-2 | 99.9% | C24H26N2O4.H3PO4.1/2H2O | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0006A
Encompass_Preferred
Carvedilol phosphate hemihydrate is a non-selective β/α-1 blocker. It inhibits lipid peroxidation with an IC50 of 5 μM and acts as a multiple-action antihypertensive agent with potential use in angina and congestive heart failure. This compound also induces autophagy and inhibits the NLRP3 inflammasome. It is soluble in DMSO at concentrations of 100 mg/mL or greater (194.75 mM) in vitro. This product is for research use only.
  • Non-selective β/α-1 blocker
  • Inhibits lipid peroxidation (IC50 of 5 μM)
  • Multiple-action antihypertensive agent
  • Potential for use in angina and congestive heart failure
  • Autophagy inducer
  • Inhibits the NLRP3 inflammasome
  • Inhibits superoxide generation by activated human neutrophils (IC50 of 28 μM)
  • Scavenges oxygen free radicals in a cell-free system (IC50 of 25 μM)
  • Purity of 99.9%
Catalog No. 50-003-59030