Learn More
Medchemexpress LLC Carvedilol phosphate hemihydrate | 610309-89-2 | 99.9% | C24H26N2O4.H3PO4.1/2H2O | 500 MG
Supplier: Medchemexpress LLC HYB0006A500MG
Carvedilol phosphate hemihydrate is a non-selective β/α-1 blocker. This multiple action antihypertensive agent has potential use in angina and congestive heart failure.
- Inhibits lipid peroxidation with an IC50 of 5 μM.
- Autophagy inducer.
- Inhibits the NLRP3 inflammasome.