Learn More
Medchemexpress LLC (-)-carvone | 6485-40-1 | MFCD00001578 | 99.9% | 150.22 g/mol | C10H14O | 1 ML

Supplier: Medchemexpress LLC HYW01742310MM/1ML
(-)-Carvone (CAS 6485-40-1) is a naturally occurring monoterpenoid that acts as an insect neurotoxin and irreversible acetylcholinesterase inhibitor. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for research applications such as AChE inhibition assays and insecticidal studies.
- Ready-to-use 10 mM solution in DMSO for convenient dosing.
- High purity (99.91%) suitable for research-grade assays.
- Molecular formula C10H14O; molecular weight 150.22 g/mol.
- Solubility and formulation guidance available for in vitro and in vivo use.
- Storage recommendations provided to maintain stock and pure-form stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.