Learn More
Medchemexpress LLC Caryophyllene oxide | 1139-30-6 | 1 ML

Supplier: Medchemexpress LLC HYN354410MM/1ML
Caryophyllene oxide, also known as (-)-Caryophyllene oxide, is a bicyclic sesquiterpene. It demonstrates anticancer effects, inducing apoptosis in PC-3 cells, and exhibits analgesic and anti-inflammatory activities. Additionally, this compound possesses insecticidal, antioxidant, antimicrobial, antifungal, and antiparasitic properties.
- Induces apoptosis of PC-3 cells
- Exhibits analgesic effects
- Anti-inflammatory activities
- Insecticidal properties
- Antioxidant properties
- Antimicrobial and antifungal activities
- Antiparasitic properties
- High purity of 99.71%
- Molecular formula: C15H24O
- Molecular weight: 220.35
- White to off-white solid appearance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.