Learn More
MOLECULAR DEPOT LLC CAS 216393-53-2 - 5 mg
Supplier:  MOLECULAR DEPOT LLC B2011337
Encompass_Preferred
N-(Biotinoyl)-N -(iodoacetyl)ethylenediamine is a thiol-reactive biotinylating reagent for proteins and other biomolecules This product is provided as a powder and has a MW of 454 3 g/mol It has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13416