Learn More
Sigma Aldrich Fine Chemicals Biosciences CAS9 Blasticidin Lenti Par1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences LVCAS9BST1EA
Encompass
Ready-to-use Cas9 lentiviral particles are ideal for transducing a wide range of cell lines. The Cas9 Blasticidin Lentiviral particles efficiently integrate Cas9 into the genomes of both dividing and non-dividing cell lines for use with CRISPR genome editing. Stably integrate Cas9 and Blast using the convenient ready-to-use lentiviral particles while avoiding the tedious lentivirus production process.
Catalog No. 50-290-4457