Learn More
Cell Biolabs Inc Cas9 ELISA Kit, 96 assays
Supplier:  Cell Biolabs Inc PRB5079
Encompass_Preferred
Cas9 is an RNA-guided DNA endonuclease that is a key part of the CRISPR adaptive immunity system. Cell Biolabs' Cas9 ELISA Kit provides a convenient method for measuring S. pyogenes Cas9 levels in a standard 96-well microplate reader. Cas9 levels as low as 1.5 ng/mL can be detected in cell or tissue lysates. Each kit provides sufficient reagents for 96 assays including standaards and unknown samples.
Catalog No. NC1748388