Learn More
BPS Bioscience Inc Cas9 Lentiviruses (Inducible Tet-On)
SDP
Supplier:  BPS Bioscience Inc 78794
Encompass
Cas9 Lentiviruses (Inducible Tet-On) are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to transduce most mammalian cells, including primary and non-dividing cells. These viruses transduce cells with the Streptococcus pyogenes Cas9 gene under a tight TRE tetracycline-inducible promoter. Two vials (500 µl x 2)
Catalog No. NC3084213