Learn More
Pna Bio CAS9 PROTEIN W/NLS 250UG
SDP
Supplier:  Pna Bio CP02
Encompass
High Concentration Cas9 Protein From Streptococcus Pyogenes With Nls (Injection Ready); Reconstitutes To 5 Mg/Ml (~30 Um); Recommended For Electroporation. 250 Ug (1560 Pmol / 30 Um X 50 Ul). Injection, Transfection Ready. Activity Validated By In Vitro Cleavage Assay, Electroporation (As Rnp Complex), And Mouse Embryo Injection For Each Batch.
Catalog No. NC0974724