Learn More
Lakeside Manufacturing Inc Closed Case Cart, Two Door, Wire Shelves, 36-In Shelf Width
Supplier:  Lakeside Manufacturing Inc 6935
Encompass_Preferred
The Lakeside 6935 Closed Case Cart has two doors with wire shelving that has a 36" shelf width. This unit is an all-welded stainless steel construction. The hold open latch secures the door open for easy cleaning. Door latch keeps contents safe and sterile (locking model available). The extended push handle offers better control. This model has wire pull-out shelves equipped with a locking mechanism. The innovative floor design directs water to the drain area. Made in the USA.
Catalog No. NC2670018
Only null left
Add to Cart
Request a Quote