Learn More
Research Products International Corp Casein, 500 ML
SDP
Supplier:  Research Products International Corp C44000500.0
Encompass_Preferred
Contains 3% Casein in TBS buffer. Optimized for use in western blotting to minimize non-specific binding of antibodies.
Catalog No. 50-197-7942