Learn More
Medchemexpress LLC Casein | 9000-71-9 | 97.4% | 2061.96 | C81H125N22O39P | 10G

Supplier: Medchemexpress LLC HY12586510G
Casein is a phosphoprotein supplied as an off-white powder for laboratory and research use. It serves as a biochemical substrate for kinases and proteases, and is used in assays, formulations, and material applications such as coatings and adhesives. Composition data include molecular weight 2061.96, formula C81H125N22O39P, and reported purity 97.4%.
- Off-white powder suitable for laboratory use.
- Acts as a substrate for kinases and proteases.
- Useful in biochemical assays and protein studies.
- Applicable in coatings, adhesives, and material formulations.
- Molecular weight 2061.96 and formula C81H125N22O39P.
- Purity reported as 97.4%.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.