Learn More
Medchemexpress LLC Casein | 9000-71-9 | 97.4% | 2061.96 g/mol | C81H125N22O39P | 50G
SDP
Supplier:  Medchemexpress LLC HY12586550G
Encompass_Preferred
Casein is a milk-derived phosphoprotein provided as a research reagent for biochemical and cell-signaling studies. It functions as a substrate in kinase assays and as an endogenous metabolite reference. Typical analytical information lists a purity of 97.4% and a molecular weight of 2061.96 g/mol; solubility and lot-specific details are available in the COA and datasheet.
  • Suitable substrate for kinase assays and enzymology studies.
  • High analytical purity for reproducible experimental results.
  • Soluble in 0.1 M NaOH at 20 mg/mL with ultrasonic warming and heating to 60°C.
  • Available in multiple laboratory pack sizes for workflow flexibility.
  • Supporting documentation includes data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-35598