Learn More
TARGETMOL CHEMICALS INC CASEIN KINASE II INHIBIT 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T106875MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Casein kinase II inhibitor IV is an effective small molecule inducer which can be used to induce epidermal keratinocyte differentiation. purity: 99%
Catalog No. 50-258-1691