Learn More
Medchemexpress LLC Casein kinase inhibitor A51 | 2079068-74-7 | 98.6% | 360.88 g/mol | C18H25ClN6 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12395425MG
Encompass_Preferred
BTX-A51 is a small-molecule, orally active inhibitor of casein kinase 1α (CK1α) reported to induce leukemia cell apoptosis and demonstrate anti-leukemic activity. It is supplied as a research-grade solid in pre-weighed quantities for laboratory use.
  • Potent, orally active inhibitor of casein kinase 1α.
  • Demonstrated induction of leukemia cell apoptosis in reported studies.
  • Supplied as a research-grade solid suitable for laboratory experiments.
  • Available in multiple pre-weighed pack sizes, including 25 mg.
  • High purity (98.6%) as provided by the supplier.
  • Molecular formula C18H25ClN6; molecular weight 360.88 g/mol.
Catalog No. 50-002-35979