Learn More
Medchemexpress LLC Casein kinase inhibitor A86 | 2079069-01-3 | 99.3% | 344.43 | C18H25FN6 | 25MG

Supplier: Medchemexpress LLC HY12395525MG
Casein Kinase inhibitor A86 is a potent, orally active casein kinase 1α (CK1α) inhibitor that also inhibits CDK7 and CDK9. It induces apoptosis in leukemia cells and is provided as an off-white to light yellow solid for research use only.
- Potent CK1α inhibitor with activity against CDK7 and CDK9.
- High purity: 99.3%.
- Molecular weight: 344.43 g/mol; formula: C18H25FN6.
- Soluble in DMSO (≈12.5 mg/mL); in vivo formulation protocols available.
- Storage: powder at -20°C for long term; in solvent at -80°C for months.
- Available in multiple small-scale packages; larger quantities available by quote.
- Supplied for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.