Learn More
ABclonal Technology Caspase-12 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2286420UL
Encompass_Preferred
Caspases are cysteine proteases that cleave C-terminal aspartic acid residues on their substrate molecules This gene is most highly related to members of the ICE subfamily of caspases that process inflammatory cytokines In rodents the homolog of this gene mediates apoptosis in response to endoplasmic reticulum stress However in humans this gene contains a polymorphism for the presence or absence of a premature stop codon The majority of human individuals have the premature stop codon and produce a truncated non-functional protein The read-through codon occurs primarily in individuals of African descent and carriers have endotoxin hypo-responsiveness and an increased susceptibility to severe sepsis Several alternatively spliced transcript variants have been noted for this gene
Catalog No. 50-000-06646