Learn More
CPC Scientific MCA-Val-Asp-Gln-Met-Asp-Gly-Trp-Lys(DNP)-NH2 0.5MG
SDP
Supplier:  CPC Scientific CASP060A
Encompass
Fluorogenic resonance energy transfer substrate for the detection of caspase-3 activity.SEQUENCE: MCA-Val-Asp-Gln-Met-Asp-Gly-Trp-Lys(DNP)-NH2ONE-LETTER SEQUENCE: Mca-VDQMDGWK-(DNP)-NH2MOLECULAR FORMULA: C60H75N14O21S1MOLECULAR WEIGHT: 1360.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:
SKU(s): CASP-060A, CASP-060B
Catalog No. 50-210-9025