Learn More
CPC Scientific Ac-Leu-Glu-His-Asp-AMC 1MG
SDP
Supplier:  CPC Scientific CASP081A
Encompass
ONE-LETTER SEQUENCE: Ac-LEHD-AMCMOLECULAR FORMULA: C33H41N7O11MOLECULAR WEIGHT:711.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [292633-16-0], RESEARCH AREA: Apoptosis
Catalog No. 50-210-9072