Learn More
Enzo Life Sciences Caspase Assay Buffer (20 ml)
SDP
Supplier:  Enzo Life Sciences BMLKI1110020
Encompass_Preferred
50 mM HEPES, pH 7.4, 100 mM NaCl, 0.1% CHAPS, 10 mM DTT, 1 mM EDTA, 10% glycerol. Shipping: Shipped on Blue Ice. Long Term Storage: -80°C
Catalog No. NC1128408