Learn More
Cayman Chemical Caspase1 InhIbItr Screeng 384
SDP
Supplier:  Cayman Chemical 701840384 WELL
Encompass_Preferred
A GPR18 agonist selective for GPR18 over GPR55 inBarrestin recruitment assays using CHO-K1 cells expressing the human receptors (EC50s 0.0454 and >10 UM respectively) does not bind to CB1 or CB2 receptors (Kis >10 UM for both)
Catalog No. 50-289-5817