Learn More
Selleck Chemical LLC Caspofungin Acetate 5mg 179463-17-3 MK-0991
SDP
Supplier:  Selleck Chemical LLC S30735MG
Encompass_Preferred
Caspofungin acetate (MK-0991) is an lipopeptide antifungal ?-1,3-glucan synthase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3357