Learn More
Medchemexpress LLC Caspofungin-d4 (MK-0991-d4; L-743872-d4) | 1131958-73-0 | 1097.34 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY17006S1MG
Encompass_Preferred
Caspofungin-d4 (MK-0991-d4; L-743872-d4) is a deuterium labeled Caspofungin, a potent antifungal agent that inhibits the synthesis of the fungal cell wall component β-(l,3)-D-glucan. This compound is valuable for research, particularly in understanding pharmacokinetic and metabolic profiles through deuteration.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
Catalog No. 50-003-91253