Learn More
Medchemexpress LLC HY-17006 50mg , Caspofungin (diacetate) CAS:179463-17-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1700650MG
Encompass_Preferred
HY-17006 50mg Caspofungin (diacetate) CAS: 179463-17-3 Caspofungin (MK-0991) diacetate is a potent antifungal agent. Caspofungin diacetate inhibits the synthesis of the fungal cell wall component β-(l,3)-D-glucan. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9772