Learn More
Electron Microscopy Sciences ActivFlo Mini Biopsy Cassettes, White
SDP
Supplier:  Electron Microscopy Sciences 70076128
Encompass_Preferred
Activflo Mini Cassettes are one third the size of standard cassettes, and feature sloped walls, creating a basket for small tissue management. The smaller size allows double the number of blocks to be stored, reducing specimen storage space. Excellent solution for microwave and rapid tissue processors.
Color: White
Catalog No. 50-192-6957