Learn More
Med Vet International Medical Tape 3M Micropore S Blue 2 inch X 5-1/2 Yard Silicone Nonsterile, 2770-2, 60/Case
Supplier:  Med Vet International 774628CS
Encompass_Preferred
Offers short-term and long-term (48 hour) reliable and pliable fixation Comfort: Removes cleanly, without disrupting fragile skin layers or causing patients undue pain Ease of use: Can be repositioned and neatly torn by hand Helps reduce risk: Individually packaged single-use rolls can help protect patients by reducing the risk of cross-contamination
Catalog No. 50-283-1753