Learn More
Uline Casters for 60" and 72" Packing Stations - Set of 4
Supplier:  Uline H7003
Encompass
Add mobility to your packing stations and workstations.5" locking swivel casters add 7" to height.60" and 72" long stations require 4 casters.
Catalog No. NC2830144