Learn More
Medchemexpress LLC Casticin (Vitexicarpin) | 479-91-4 | 374.34 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0516
Encompass_Preferred
Casticin is a methoxylated flavonol isolated from *Vitex rotundifolia* with antimitotic and anti-inflammatory effects. It inhibits the activation of STAT3, a protein involved in cell growth and division.
  • Inhibits proliferation of KB cells and LX2 cells.
  • Induces apoptosis in L02 cells.
  • Reduces viability of 786-O, YD-8, and HN-9 cells.
  • Increases cleavage caspase-3 and PPAR.
  • Diminishes levels of Bcl-xl, Bcl-2, IAP-1/-2, VEGF, MMP-9, and COX-2 proteins.
  • Promotes apoptotic cell death in tumor cells.
  • Enhances the effect of ionizing radiation in 786-O cells.
  • Attenuates liver fibrosis induced by CCl4 or BDL in vivo.
Catalog No. 50-003-90243