Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Casuarinin | 79786-01-9 | 936.65 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN127171MG

Encompass_Preferred

Casuarinin is an active product that can be isolated from the bark of *Terminalia arjuna*. It induces apoptosis and cell cycle arrest.

  • Purity: 98.24%
  • Appearance: Solid, light yellow to light brown
  • Storage: -20°C, sealed storage, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).
  • For research use only.

Catalog No. 50-004-16502


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.