Learn More
STOGGLES Cat Eye Peach Dimmers
Supplier:  STOGGLES 134
Encompass_Preferred
Our Dimmers Lens are a UV-responsive lens that transition from light to dark based off of light exposure. Our light responsive lenses are anti-fog coated and shatter proof ensuring superior clarity and quality.
Catalog No. NC2388259