Learn More
Biocare Medical, LLC CAT HEMATOXYLIN 1GAL
SDP
Supplier:  Biocare Medical, LLC CATHEGL
Encompass_Preferred
CAT Hematoxylin is a modified Lillie-Mayer's Method formulation that provides incredible nuclear detail in routine H&E's and IHC, ard some special stains. It is formulated to eliminate the necessity for section differentiation.
Catalog No. 50-823-91