Learn More
Grainger Cat5e Shielded Inline Coupler
Supplier:  Grainger 14J353
Encompass_Preferred
The Monoprice Shielded Inline Coupler is a BNC connector that joins two coaxial cables together while maintaining the shielding between them. It has a 50 ohm impedance and is designed for CCTV or other video applications using RG59 or RG6 coax cable. The nickel-plated center pin and connector body provide corrosion resistance. This inline coupler allows you to extend your existing coax cable run when you need a little more length. It's a cost-effective solution for splicing cables together without signal loss.
Catalog No. 50-275-2753