Learn More
Biomatik Corporation FCGR2B anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07640
Encompass_Preferred
The FCGR2B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Low affinity immunoglobulin gamma Fc region receptor II-b protein (12-289AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Low affinity immunoglobulin gamma Fc region receptor II-bIgG Fc receptor II-b, CDw32, Fc-gamma RII-b, Fc-gamma-RIIb, FcRII-b, CD32, FCGR2B, CD32, FCG2, IGFR2. UniProt: P31994. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7225